Anti-KRT20 Mouse Monoclonal Antibody [clone: 11A1]

Proveedor: OriGene
CF802582
ORIGCF802582EA 764 EUR
ORIGCF802582
Anti-KRT20 Mouse Monoclonal Antibody [clone: 11A1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: KRT20
Clonality: Monoclonal
Clone: 11A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR