Anti-TOP2A Mouse Monoclonal Antibody [clone: 2H9]

Proveedor: OriGene
CF802573
ORIGCF802573EA 764 EUR
ORIGCF802573
Anti-TOP2A Mouse Monoclonal Antibody [clone: 2H9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TOP2A
Clonality: Monoclonal
Clone: 2H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR