Anti-PARN Mouse Monoclonal Antibody [clone: 4D6]

Proveedor: OriGene
CF802569
ORIGCF802569EA 764 EUR
ORIGCF802569
Anti-PARN Mouse Monoclonal Antibody [clone: 4D6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PARN
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR