Anti-STK11 Mouse Monoclonal Antibody [clone: 10A3]

Proveedor: OriGene
CF802554
ORIGCF802554EA 764 EUR
ORIGCF802554
Anti-STK11 Mouse Monoclonal Antibody [clone: 10A3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: STK11
Clonality: Monoclonal
Clone: 10A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR