Anti-KLK8 Mouse Monoclonal Antibody [clone: 5B4]

Proveedor: OriGene
CF506159
ORIGCF506159EA 764 EUR
ORIGCF506159
Anti-KLK8 Mouse Monoclonal Antibody [clone: 5B4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: KLK8
Clonality: Monoclonal
Clone: 5B4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR