Anti-ZSCAN21 Mouse Monoclonal Antibody [clone: 3D4]

Proveedor: OriGene
ORIGCF506147EA 764 EUR
ORIGCF506147
Anti-ZSCAN21 Mouse Monoclonal Antibody [clone: 3D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: ZSCAN21
Clonality: Monoclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR