Anti-ENG Mouse Monoclonal Antibody [clone: 6G8]

Proveedor: OriGene
CF506144
ORIGCF506144EA 764 EUR
ORIGCF506144
Anti-ENG Mouse Monoclonal Antibody [clone: 6G8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: 6G8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR