Anti-MS4A1 Mouse Monoclonal Antibody [clone: 2D3]

Proveedor: OriGene
CF800487
ORIGCF800487EA 764 EUR
ORIGCF800487
Anti-MS4A1 Mouse Monoclonal Antibody [clone: 2D3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:500

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR