Anti-ZFAND5 Mouse Monoclonal Antibody [clone: 1D8]

Proveedor: OriGene
CF800482
ORIGCF800482EA 764 EUR
ORIGCF800482
Anti-ZFAND5 Mouse Monoclonal Antibody [clone: 1D8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: ZFAND5
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR