Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 1B1]

Proveedor: OriGene
ORIGCF506905EA 786 EUR
ORIGCF506905
Anti-MADCAM1 Mouse Monoclonal Antibody [clone: 1B1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000

Type: Primary
Antigen: MADCAM1
Clonality: Monoclonal
Clone: 1B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR