Anti-ICAM1 Mouse Monoclonal Antibody [clone: 2H4]

Proveedor: OriGene
CF506870
ORIGCF506870EA 764 EUR
ORIGCF506870
Anti-ICAM1 Mouse Monoclonal Antibody [clone: 2H4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 2H4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR