Anti-DOT1L Mouse Monoclonal Antibody [clone: 1D8]

Proveedor: OriGene
CF802482
ORIGCF802482EA 764 EUR
ORIGCF802482
Anti-DOT1L Mouse Monoclonal Antibody [clone: 1D8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: DOT1L
Clonality: Monoclonal
Clone: 1D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR