Anti-OTC Mouse Monoclonal Antibody [clone: 4G4]

Proveedor: OriGene
CF802454
ORIGCF802454EA 786 EUR
ORIGCF802454
Anti-OTC Mouse Monoclonal Antibody [clone: 4G4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150

Type: Primary
Antigen: OTC
Clonality: Monoclonal
Clone: 4G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR