Anti-KI67 Mouse Monoclonal Antibody [clone: 13B11]

Proveedor: OriGene
CF802432
ORIGCF802432EA 764 EUR
ORIGCF802432
Anti-KI67 Mouse Monoclonal Antibody [clone: 13B11]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100-500

Type: Primary
Antigen: KI67
Clonality: Monoclonal
Clone: 13B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR