Anti-KI67 Mouse Monoclonal Antibody [clone: 15A4]

Proveedor: OriGene
CF802429
ORIGCF802429EA 764 EUR
ORIGCF802429
Anti-KI67 Mouse Monoclonal Antibody [clone: 15A4]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:100; Western Blot: 1:200-1:1000

Type: Primary
Antigen: KI67
Clonality: Monoclonal
Clone: 15A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR