Anti-DNM1L Mouse Monoclonal Antibody [clone: 1C1]

Proveedor: OriGene
CF506116
ORIGCF506116EA 764 EUR
ORIGCF506116
Anti-DNM1L Mouse Monoclonal Antibody [clone: 1C1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: DNM1L
Clonality: Monoclonal
Clone: 1C1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR