Anti-SYP Mouse Monoclonal Antibody [clone: 1A1]

Proveedor: OriGene
CF506105
ORIGCF506105EA 764 EUR
ORIGCF506105
Anti-SYP Mouse Monoclonal Antibody [clone: 1A1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: SYP
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR