Anti-PTPRC Mouse Monoclonal Antibody [clone: 3D8]

Proveedor: OriGene
CF506104
ORIGCF506104EA 764 EUR
ORIGCF506104
Anti-PTPRC Mouse Monoclonal Antibody [clone: 3D8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 3D8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR