Anti-CHGA Mouse Monoclonal Antibody [clone: 2E12]

Proveedor: OriGene
CF506098
ORIGCF506098EA 786 EUR
ORIGCF506098
Anti-CHGA Mouse Monoclonal Antibody [clone: 2E12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: 2E12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR