Anti-MS4A1 Mouse Monoclonal Antibody [clone: 1C12]

Proveedor: OriGene
ORIGCF800464EA 764 EUR
ORIGCF800464
Anti-MS4A1 Mouse Monoclonal Antibody [clone: 1C12]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR