Anti-BCL10 Mouse Monoclonal Antibody [clone: 2G9]

Proveedor: OriGene
CF800455
ORIGCF800455EA 764 EUR
ORIGCF800455
Anti-BCL10 Mouse Monoclonal Antibody [clone: 2G9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: BCL10
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR