Anti-MTIF3 Mouse Monoclonal Antibody [clone: 4G2]

Proveedor: OriGene
CF800420
ORIGCF800420EA 786 EUR
ORIGCF800420
Anti-MTIF3 Mouse Monoclonal Antibody [clone: 4G2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: MTIF3
Clonality: Monoclonal
Clone: 4G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR