Anti-CD44 Mouse Monoclonal Antibody [clone: 2E3]

Proveedor: OriGene
CF506834
ORIGCF506834EA 764 EUR
ORIGCF506834
Anti-CD44 Mouse Monoclonal Antibody [clone: 2E3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 2E3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR