Anti-SIVA1 Mouse Monoclonal Antibody [clone: 2F8]

Proveedor: OriGene
CF506830
ORIGCF506830EA 764 EUR
ORIGCF506830
Anti-SIVA1 Mouse Monoclonal Antibody [clone: 2F8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SIVA1
Clonality: Monoclonal
Clone: 2F8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR