Anti-CD5 Mouse Monoclonal Antibody [clone: 4F9]

Proveedor: OriGene
CF802411
ORIGCF802411EA 764 EUR
ORIGCF802411
Anti-CD5 Mouse Monoclonal Antibody [clone: 4F9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:0

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 4F9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR