Anti-CD5 Mouse Monoclonal Antibody [clone: 1B7]

Proveedor: OriGene
CF802402
ORIGCF802402EA 764 EUR
ORIGCF802402
Anti-CD5 Mouse Monoclonal Antibody [clone: 1B7]
Anticuerpos
Recommended Dilutions: Flow Cytometry: 1:50

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR