Anti-SOX9 Mouse Monoclonal Antibody [clone: 3G3]

Proveedor: OriGene
CF802392
ORIGCF802392EA 764 EUR
ORIGCF802392
Anti-SOX9 Mouse Monoclonal Antibody [clone: 3G3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SOX9
Clonality: Monoclonal
Clone: 3G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR