Anti-TOP2A Mouse Monoclonal Antibody [clone: 1D4]

Proveedor: OriGene
CF802383
ORIGCF802383EA 764 EUR
ORIGCF802383
Anti-TOP2A Mouse Monoclonal Antibody [clone: 1D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: TOP2A
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR