Anti-RARRES1 Mouse Monoclonal Antibody [clone: 1B8]

Proveedor: OriGene
CF506091
ORIGCF506091EA 764 EUR
ORIGCF506091
Anti-RARRES1 Mouse Monoclonal Antibody [clone: 1B8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunofluorescence: 1:100

Type: Primary
Antigen: RARRES1
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR