Anti-RARRES1 Mouse Monoclonal Antibody [clone: 1D2]

Proveedor: OriGene
CF506089
ORIGCF506089EA 764 EUR
ORIGCF506089
Anti-RARRES1 Mouse Monoclonal Antibody [clone: 1D2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: RARRES1
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR