Anti-PTPRC Mouse Monoclonal Antibody [clone: 1B7]

Proveedor: OriGene
CF506054
ORIGCF506054EA 764 EUR
ORIGCF506054
Anti-PTPRC Mouse Monoclonal Antibody [clone: 1B7]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-8000; Immunochemistry: 1:150; Flow Cytometry: 1:50

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR