Anti-EPHX1 Mouse Monoclonal Antibody [clone: 2C1]

Proveedor: OriGene
CF800408
ORIGCF800408EA 764 EUR
ORIGCF800408
Anti-EPHX1 Mouse Monoclonal Antibody [clone: 2C1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: EPHX1
Clonality: Monoclonal
Clone: 2C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR