Anti-EPHX1 Mouse Monoclonal Antibody [clone: 3A10]

Proveedor: OriGene
CF800395
ORIGCF800395EA 764 EUR
ORIGCF800395
Anti-EPHX1 Mouse Monoclonal Antibody [clone: 3A10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: EPHX1
Clonality: Monoclonal
Clone: 3A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR