Anti-MORF4 Mouse Monoclonal Antibody [clone: 5A6]

Proveedor: OriGene
CF800376
ORIGCF800376EA 764 EUR
ORIGCF800376
Anti-MORF4 Mouse Monoclonal Antibody [clone: 5A6]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MORF4
Clonality: Monoclonal
Clone: 5A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR