Anti-C11orf67 Mouse Monoclonal Antibody [clone: 4C1]

Proveedor: OriGene
ORIGCF803536EA 764 EUR
ORIGCF803536
Anti-C11orf67 Mouse Monoclonal Antibody [clone: 4C1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: C11orf67
Clonality: Monoclonal
Clone: 4C1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR