Anti-VEGFA Mouse Monoclonal Antibody [clone: 2B9]

Proveedor: OriGene
CF802346
ORIGCF802346EA 764 EUR
ORIGCF802346
Anti-VEGFA Mouse Monoclonal Antibody [clone: 2B9]
Anticuerpos
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: VEGFA
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR