Anti-ZEB1 Mouse Monoclonal Antibody [clone: 8E2]

Proveedor: OriGene
CF802295
ORIGCF802295EA 764 EUR
ORIGCF802295
Anti-ZEB1 Mouse Monoclonal Antibody [clone: 8E2]
Anticuerpos
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:150

Type: Primary
Antigen: ZEB1
Clonality: Monoclonal
Clone: 8E2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR