Anti-SPATA2L Mouse Monoclonal Antibody [clone: 2B4]

Proveedor: OriGene
ORIGCF802370EA 764 EUR
ORIGCF802370
Anti-SPATA2L Mouse Monoclonal Antibody [clone: 2B4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: SPATA2L
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR