Anti-KLK8 Mouse Monoclonal Antibody [clone: 1G8]

Proveedor: OriGene
CF801596
ORIGCF801596EA 764 EUR
ORIGCF801596
Anti-KLK8 Mouse Monoclonal Antibody [clone: 1G8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: KLK8
Clonality: Monoclonal
Clone: 1G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR