Anti-WWC1 Mouse Monoclonal Antibody [clone: 1H8]

Proveedor: OriGene
CF801568
ORIGCF801568EA 764 EUR
ORIGCF801568
Anti-WWC1 Mouse Monoclonal Antibody [clone: 1H8]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: WWC1
Clonality: Monoclonal
Clone: 1H8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR