Anti-RASGRP3 Mouse Monoclonal Antibody [clone: 5C5]

Proveedor: OriGene
CF801527
ORIGCF801527EA 786 EUR
ORIGCF801527
Anti-RASGRP3 Mouse Monoclonal Antibody [clone: 5C5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: RASGRP3
Clonality: Monoclonal
Clone: 5C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR