Anti-ARL2BP Mouse Monoclonal Antibody [clone: 1A10]

Proveedor: OriGene
CF800297
ORIGCF800297EA 764 EUR
ORIGCF800297
Anti-ARL2BP Mouse Monoclonal Antibody [clone: 1A10]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ARL2BP
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR