Anti-VWF Mouse Monoclonal Antibody [clone: 2D11]

Proveedor: OriGene
CF803447
ORIGCF803447EA 764 EUR
ORIGCF803447
Anti-VWF Mouse Monoclonal Antibody [clone: 2D11]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR