Anti-PTGES3 Mouse Monoclonal Antibody [clone: 4B3]

Proveedor: OriGene
CF803437
ORIGCF803437EA 764 EUR
ORIGCF803437
Anti-PTGES3 Mouse Monoclonal Antibody [clone: 4B3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: PTGES3
Clonality: Monoclonal
Clone: 4B3
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR