Anti-MAP1LC3A Mouse Monoclonal Antibody [clone: 4C5]

Proveedor: OriGene
CF803408
ORIGCF803408EA 764 EUR
ORIGCF803408
Anti-MAP1LC3A Mouse Monoclonal Antibody [clone: 4C5]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: MAP1LC3A
Clonality: Monoclonal
Clone: 4C5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR