Anti-CD63 Mouse Monoclonal Antibody [clone: 2A2E2]

Proveedor: OriGene
CF803394
ORIGCF803394EA 764 EUR
ORIGCF803394
Anti-CD63 Mouse Monoclonal Antibody [clone: 2A2E2]
Anticuerpos
Recommended Dilutions: Immunochemistry: 1:150; Flow Cytometry: 1:50

Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: 2A2E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR