Anti-DDIT3 Mouse Monoclonal Antibody [clone: 4C4]

Proveedor: OriGene
CF802207
ORIGCF802207EA 764 EUR
ORIGCF802207
Anti-DDIT3 Mouse Monoclonal Antibody [clone: 4C4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: DDIT3
Clonality: Monoclonal
Clone: 4C4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR