Anti-KLK8 Mouse Monoclonal Antibody [clone: 3F3]

Proveedor: OriGene
CF801460
ORIGCF801460EA 764 EUR
ORIGCF801460
Anti-KLK8 Mouse Monoclonal Antibody [clone: 3F3]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: KLK8
Clonality: Monoclonal
Clone: 3F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR