Anti-MAFB Mouse Monoclonal Antibody [clone: 1F1]

Proveedor: OriGene
CF800279
ORIGCF800279EA 764 EUR
ORIGCF800279
Anti-MAFB Mouse Monoclonal Antibody [clone: 1F1]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: MAFB
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR