Anti-VWF Mouse Monoclonal Antibody [clone: 1D4]

Proveedor: OriGene
CF803310
ORIGCF803310EA 764 EUR
ORIGCF803310
Anti-VWF Mouse Monoclonal Antibody [clone: 1D4]
Anticuerpos
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR